missing translation for 'onlineSavingsMsg'
Learn More

Dimethylamine, 40% w/w Aqueous, Reagents

Catalog No. p-200021851
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336510 1 L
01336509 2.5 L
01336508 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01336510 Supplier Labchem Inc Supplier No. C71960001C
Only null left

Dimethylamine, 40% w/w Aqueous, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 124-40-3
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym N-Methylmethanamine
IUPAC Name water
SMILES O

Specifications

Color Colorless
pH 14
Packaging Glass Bottle
Quantity 1 L
Specific Gravity 0.89
Percent Purity 40%
Grade Reagent
Physical Form Liquid
Chemical Name or Material Dimethylamine, 40% w/w Aqueous