missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethylamine, 40% w/w Aqueous, Reagents
Dimethylamine, 40% w/w Aqueous, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C719600025D1
Specifications
| Dimethylamine, 40% w/w Aqueous | |
| Colorless | |
| Glass Bottle | |
| 2.5 L | |
| N-Methylmethanamine | |
| O | |
| 18.02 | |
| Reagent |
| 7732-18-5,124-40-3 | |
| 14 | |
| H2O | |
| 0.89 | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water | |
| 40% | |
| Liquid |
Chemical Identifiers
| 7732-18-5 | |
| 18.02 | |
| N-Methylmethanamine | |
| O |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water |