missing translation for 'onlineSavingsMsg'
Learn More

Dimethylamine, 26% w/v, Reagents

Catalog No. 01335416
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01335416 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01335416 Supplier Labchem Inc Supplier No. CD2611001C
Only null left

Dimethylamine, 26% w/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 124-40-3
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym N-Methylmethanamine
IUPAC Name water
SMILES O

Specifications

Color Colorless to Light Yellow
pH 10
Packaging Glass Bottle
Quantity 1 L
Specific Gravity 0.9
Percent Purity 26%
Grade Reagent
Physical Form Liquid
Chemical Name or Material Dimethylamine, 26% w/v