missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl trans-3-Hexenedioate 98.0+%, TCI America™

Catalog No. p-7130717
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H07765G 5 g
H077625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H07765G Supplier TCI America Supplier No. H07765G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 25126-93-6
Molecular Formula C8H12O4
Molecular Weight (g/mol) 172.18
MDL Number MFCD00145425
InChI Key CPSQPCGPYWRIPB-ONEGZZNKSA-N
Synonym trans-3-Hexenedioic Acid Dimethyl Ester
PubChem CID 5372575
IUPAC Name 1,6-dimethyl (3E)-hex-3-enedioate
SMILES COC(=O)C\C=C\CC(=O)OC

Specifications

Color Yellow
Boiling Point 125°C
Quantity 5 g
Formula Weight 172.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethyl trans-3-Hexenedioate