accessibility menu, dialog, popup

UserName

CAS RN 25126-93-6

IUPAC Name: 1,6-dimethyl (3E)-hex-3-enedioate
Synonyms: 1,6-dimethyl (3E)-hex-3-enedioate
Molecular Weight (g/mol): 172.18
Molecular Formula: C8H12O4
InChi Key: CPSQPCGPYWRIPB-ONEGZZNKSA-N
SMILES: COC(=O)C\C=C\CC(=O)OC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results