missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl Sulfoxide, PHOTREX™ Reagent, J.T. Baker™
High purity solvents characterized for use in UV, visible and IR spectrophotometry
$32316.78 - $32316.78
Chemical Identifiers
| CAS | 67-68-5 |
|---|---|
| Molecular Formula | C2H6OS |
| Molecular Weight (g/mol) | 78.13 |
| MDL Number | MFCD00002089 |
| InChI Key | IAZDPXIOMUYVGZ-UHFFFAOYSA-N |
| Synonym | dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide |
| PubChem CID | 679 |
| ChEBI | CHEBI:28262 |
| IUPAC Name | methanesulfinylmethane |
| SMILES | CS(C)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
02002731
|
Avantor J.T.Baker
919433 |
4 L | Narrow Mouth PVC-coated Amber Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
- For Spectrophotometry
Chemical Identifiers
| 67-68-5 | |
| 78.13 | |
| IAZDPXIOMUYVGZ-UHFFFAOYSA-N | |
| 679 | |
| methanesulfinylmethane |
| C2H6OS | |
| MFCD00002089 | |
| dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide | |
| CHEBI:28262 | |
| CS(C)=O |
Specifications
| 67-68-5 | |
| 89.00°C | |
| 1L = 1.10kg | |
| MFCD00002089 | |
| IAZDPXIOMUYVGZ-UHFFFAOYSA-N | |
| methanesulfinylmethane | |
| 679 | |
| 78.13 | |
| Dimethyl Sulfoxide |
| 4 L | |
| Narrow Mouth PVC-coated Amber Bottle | |
| C2H6OS | |
| dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide | |
| CS(C)=O | |
| 78.13 | |
| CHEBI:28262 | |
| PHOTREX™ Reagent |