missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl Sulfoxide, Multi-Compendial, J.T. Baker™
Supplier: Avantor J.T.Baker 903333
Specifications
| Dimethyl Sulfoxide | |
| 4 L | |
| Narrow Mouth PVC-coated Amber Bottle | |
| C2H6OS | |
| dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide | |
| CS(C)=O | |
| 78.13 | |
| CHEBI:28262 | |
| Multi-Compendial |
| 67-68-5 | |
| 89.00°C | |
| 1L = 1.10kg | |
| MFCD00002089 | |
| IAZDPXIOMUYVGZ-UHFFFAOYSA-N | |
| methylsulfinylmethane | |
| 679 | |
| 78.13 |
Chemical Identifiers
| 67-68-5 | |
| 78.13 | |
| IAZDPXIOMUYVGZ-UHFFFAOYSA-N | |
| 679 | |
| CS(C)=O |
| C2H6OS | |
| MFCD00002089 | |
| dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide | |
| CHEBI:28262 |