missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Sulfoxide, Multi-Compendial, J.T. Baker™

Catalog No. p-5391023
Change view
Click to view available options
Quantity:
100 mL
4 L
19 L
Packaging:
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC-coated Amber Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02002727 100 mL Narrow Mouth Amber Glass Bottle
02002729 4 L Narrow Mouth PVC-coated Amber Bottle
02002736 19 L Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 02002727 Supplier Avantor Supplier No. 903304
Only null left

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
PubChem CID 679
ChEBI CHEBI:28262
SMILES CS(C)=O

Specifications

Quantity 100 mL
Flash Point 89.00°C
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 1.10kg
IUPAC Name methylsulfinylmethane
Formula Weight 78.13
Grade Multi-Compendial
Chemical Name or Material Dimethyl Sulfoxide