missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl Sulfoxide, B&J Brand™, for HPLC, GC and spectrophotometry, >99.5%, Honeywell Burdick & Jackson
$2045.75 - $2045.75
Chemical Identifiers
| CAS | 67-68-5 |
|---|---|
| Molecular Formula | C2H6OS |
| Molecular Weight (g/mol) | 78.13 |
| MDL Number | MFCD00002089 |
| InChI Key | IAZDPXIOMUYVGZ-UHFFFAOYSA-N |
| Synonym | dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide |
| PubChem CID | 679 |
| ChEBI | CHEBI:28262 |
| IUPAC Name | methanesulfinylmethane |
| SMILES | CS(C)=O |
Chemical Identifiers
| 67-68-5 | |
| 78.13 | |
| IAZDPXIOMUYVGZ-UHFFFAOYSA-N | |
| 679 | |
| methanesulfinylmethane |
| C2H6OS | |
| MFCD00002089 | |
| dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide | |
| CHEBI:28262 | |
| CS(C)=O |
Specifications
| 67-68-5 | |
| 100.0000% | |
| 189°C | |
| 88°C | |
| C2H6OS | |
| 0.04% max. | |
| dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide | |
| IAZDPXIOMUYVGZ-UHFFFAOYSA-N | |
| methanesulfinylmethane | |
| 679 | |
| 78.13 | |
| HPLC |
| 0.5 AU max. at 275 nm (UV), 0.2 AU max. at 300 nm (UV), 0.02 AU max. at 350 nm (UV), 0.005 AU max. at 400 nm (UV) | |
| 1.0954 g/cm3 | |
| 6 x 1 L | |
| Glass Bottle | |
| MFCD00002089 | |
| 506008 | |
| Miscible in Water at 20°C | |
| CS(C)=O | |
| 78.13 | |
| CHEBI:28262 | |
| >99.5% | |
| Dimethyl Sulfoxide, B&J Brand™, for HPLC, GC and spectrophotometry, >99.5% |
Safety and Handling
ShelfLife : 547 days from date of manufacture