missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl Sulfoxide, B&J Brand™, for HPLC, GC and spectrophotometry, >99.5%, Honeywell Burdick & Jackson
Specifications
Specifications
| Absorbance | 0.5 AU max. at 275 nm (UV), 0.2 AU max. at 300 nm (UV), 0.02 AU max. at 350 nm (UV), 0.005 AU max. at 400 nm (UV) |
| CAS Max % | 100.0000% |
| Density | 1.0954 g/cm3 |
| Quantity | 6 x 1 L |
| Flash Point | 88°C |
| Packaging | Glass Bottle |
| Water | 0.04% max. |
| Beilstein | 506008 |
| Boiling Point | 189°C |
| Solubility Information | Miscible in Water at 20°C |
| Show More |
Safety and Handling
| ShelfLife | 547 days from date of manufacture |