missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl sulfoxide, ≥99.5%, Reagent Grade, Honeywell™
Reagent Grade,≥99.5%
$201.98 - $201.98
Chemical Identifiers
| CAS | 67-68-5 |
|---|---|
| Molecular Formula | C2H6OS |
| Molecular Weight (g/mol) | 78.13 |
| MDL Number | MFCD00002089 |
| InChI Key | IAZDPXIOMUYVGZ-UHFFFAOYSA-N |
| Synonym | dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide |
| PubChem CID | 679 |
| ChEBI | CHEBI:28262 |
| IUPAC Name | methanesulfinylmethane |
| SMILES | CS(C)=O |
Description
- ≥95% for general laboratory use.
Chemical Identifiers
| 67-68-5 | |
| 78.13 | |
| IAZDPXIOMUYVGZ-UHFFFAOYSA-N | |
| 679 | |
| methanesulfinylmethane |
| C2H6OS | |
| MFCD00002089 | |
| dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide | |
| CHEBI:28262 | |
| CS(C)=O |
Specifications
| 18.5°C | |
| Colorless | |
| 189°C | |
| 42% (63°F) | |
| 573°F | |
| Poly Bottle | |
| C2H6OS | |
| MFCD00002089 | |
| 506008 | |
| IAZDPXIOMUYVGZ-UHFFFAOYSA-N | |
| methanesulfinylmethane | |
| 679 | |
| 78.13g/mol | |
| Dimethyl sulfoxide |
| 67-68-5 | |
| 1.10g/mL | |
| 500 mL | |
| 87°C (188.6°F) | |
| Characteristic | |
| Liquid | |
| (CH3)2SO | |
| 0.42mmHg (20°C) | |
| dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide | |
| CS(C)=O | |
| 78.13 | |
| CHEBI:28262 | |
| ≥99.5% |
Safety and Handling
RTECSNumber : 200-664-3