missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl Suberimidate Dihydrochloride 98.0+%, TCI America™
Supplier: TCI America S024625G
Specifications
| Dimethyl Suberimidate Dihydrochloride [Cross-linking Agent for Protein Research] | |
| 221°C | |
| C10H22Cl2N2O2 | |
| 25 g | |
| ILKCDNKCNSNFMP-UHFFFAOYSA-N | |
| dimethyl octanediimidate;dihydrochloride | |
| 118696 | |
| ≥98.0% (T) |
| 34490-86-3 | |
| White | |
| MFCD00012574 | |
| dimethyl suberimidate dihydrochloride, octanediimidic acid, dimethyl ester, dihydrochloride, dimethyl octanebis imidate dihydrochloride, suberimidic acid dimethyl ester dihydrochloride, octanediimidic acid, 1,8-dimethyl ester, hydrochloride 1:2, 1,8-dimethyl octanediimidate dihydrochloride, suberimidic acid, dimethyl ester, dihydrochloride, acmc-1aiyg, 1,8-dimethoxyoctane-1,8-diyldiammonium dichloride, dimethylsuberimidate dihydrochloride | |
| COC(=N)CCCCCCC(=N)OC.Cl.Cl | |
| 273.198 | |
| 273.20 | |
| Crystalline Powder |
Chemical Identifiers
| 34490-86-3 | |
| 273.198 | |
| ILKCDNKCNSNFMP-UHFFFAOYSA-N | |
| 118696 | |
| COC(=N)CCCCCCC(=N)OC.Cl.Cl |
| C10H22Cl2N2O2 | |
| MFCD00012574 | |
| dimethyl suberimidate dihydrochloride, octanediimidic acid, dimethyl ester, dihydrochloride, dimethyl octanebis imidate dihydrochloride, suberimidic acid dimethyl ester dihydrochloride, octanediimidic acid, 1,8-dimethyl ester, hydrochloride 1:2, 1,8-dimethyl octanediimidate dihydrochloride, suberimidic acid, dimethyl ester, dihydrochloride, acmc-1aiyg, 1,8-dimethoxyoctane-1,8-diyldiammonium dichloride, dimethylsuberimidate dihydrochloride | |
| dimethyl octanediimidate;dihydrochloride |
Safety and Handling
TSCA : Yes
Recommended Storage : Refrigerator