accessibility menu, dialog, popup

UserName

HHClClH3CONHNHOCH3

CAS RN 34490-86-3

IUPAC Name: dihydrogen 1,8-dimethyl octanediimidate dichloride
Synonyms: dihydrogen 1,8-dimethyl octanediimidate dichloride
Molecular Weight (g/mol): 273.2
Molecular Formula: C10H22Cl2N2O2
InChi Key: ILKCDNKCNSNFMP-UHFFFAOYSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].COC(=N)CCCCCCC(=N)OC
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)