missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl Suberimidate Dihydrochloride 98.0+%, TCI America™
$158.44 - $525.67
Chemical Identifiers
| CAS | 34490-86-3 |
|---|---|
| Molecular Formula | C10H22Cl2N2O2 |
| Molecular Weight (g/mol) | 273.198 |
| MDL Number | MFCD00012574 |
| InChI Key | ILKCDNKCNSNFMP-UHFFFAOYSA-N |
| Synonym | dimethyl suberimidate dihydrochloride, octanediimidic acid, dimethyl ester, dihydrochloride, dimethyl octanebis imidate dihydrochloride, suberimidic acid dimethyl ester dihydrochloride, octanediimidic acid, 1,8-dimethyl ester, hydrochloride 1:2, 1,8-dimethyl octanediimidate dihydrochloride, suberimidic acid, dimethyl ester, dihydrochloride, acmc-1aiyg, 1,8-dimethoxyoctane-1,8-diyldiammonium dichloride, dimethylsuberimidate dihydrochloride |
| PubChem CID | 118696 |
| IUPAC Name | dimethyl octanediimidate;dihydrochloride |
| SMILES | COC(=N)CCCCCCC(=N)OC.Cl.Cl |
Chemical Identifiers
| 34490-86-3 | |
| 273.198 | |
| ILKCDNKCNSNFMP-UHFFFAOYSA-N | |
| 118696 | |
| COC(=N)CCCCCCC(=N)OC.Cl.Cl |
| C10H22Cl2N2O2 | |
| MFCD00012574 | |
| dimethyl suberimidate dihydrochloride, octanediimidic acid, dimethyl ester, dihydrochloride, dimethyl octanebis imidate dihydrochloride, suberimidic acid dimethyl ester dihydrochloride, octanediimidic acid, 1,8-dimethyl ester, hydrochloride 1:2, 1,8-dimethyl octanediimidate dihydrochloride, suberimidic acid, dimethyl ester, dihydrochloride, acmc-1aiyg, 1,8-dimethoxyoctane-1,8-diyldiammonium dichloride, dimethylsuberimidate dihydrochloride | |
| dimethyl octanediimidate;dihydrochloride |
Specifications
| 34490-86-3 | |
| White | |
| MFCD00012574 | |
| dimethyl suberimidate dihydrochloride, octanediimidic acid, dimethyl ester, dihydrochloride, dimethyl octanebis imidate dihydrochloride, suberimidic acid dimethyl ester dihydrochloride, octanediimidic acid, 1,8-dimethyl ester, hydrochloride 1:2, 1,8-dimethyl octanediimidate dihydrochloride, suberimidic acid, dimethyl ester, dihydrochloride, acmc-1aiyg, 1,8-dimethoxyoctane-1,8-diyldiammonium dichloride, dimethylsuberimidate dihydrochloride | |
| COC(=N)CCCCCCC(=N)OC.Cl.Cl | |
| 273.198 | |
| 273.20 | |
| Crystalline Powder |
| 221°C | |
| C10H22Cl2N2O2 | |
| 5 g | |
| ILKCDNKCNSNFMP-UHFFFAOYSA-N | |
| dimethyl octanediimidate;dihydrochloride | |
| 118696 | |
| ≥98.0% (T) | |
| Dimethyl Suberimidate Dihydrochloride [Cross-linking Agent for Protein Research] |
Safety and Handling
TSCA : Yes
Recommended Storage : Refrigerator