missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl Suberate 98.0+%, TCI America™
$290.79 - $3441.05
Chemical Identifiers
| CAS | 1732-09-8 |
|---|---|
| Molecular Formula | C10H18O4 |
| Molecular Weight (g/mol) | 202.25 |
| MDL Number | MFCD00008471 |
| InChI Key | LNLCRJXCNQABMV-UHFFFAOYSA-N |
| Synonym | dimethyl suberate, octanedioic acid, dimethyl ester, suberic acid, dimethyl ester, octanedioic acid dimethyl ester, suberic acid dimethyl ester, octanedioic acid dimethyl, 1,8-dimethyl octanedioate, octanedioic acid, 1,8-dimethyl ester, dimethyl octane-1,8-dioate |
| PubChem CID | 15611 |
| ChEBI | CHEBI:81345 |
| IUPAC Name | 1,8-dimethyl octanedioate |
| SMILES | COC(=O)CCCCCCC(=O)OC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
O018325ML
|
TCI America
O018325ML |
25 mL |
Each for $290.79
|
|
|||||
|
O0183500ML
|
TCI America
O0183500ML |
500 mL |
Each for $3,441.05
|
|
|||||
Chemical Identifiers
| 1732-09-8 | |
| 202.25 | |
| LNLCRJXCNQABMV-UHFFFAOYSA-N | |
| 15611 | |
| 1,8-dimethyl octanedioate |
| C10H18O4 | |
| MFCD00008471 | |
| dimethyl suberate, octanedioic acid, dimethyl ester, suberic acid, dimethyl ester, octanedioic acid dimethyl ester, suberic acid dimethyl ester, octanedioic acid dimethyl, 1,8-dimethyl octanedioate, octanedioic acid, 1,8-dimethyl ester, dimethyl octane-1,8-dioate | |
| CHEBI:81345 | |
| COC(=O)CCCCCCC(=O)OC |
Specifications
| 1732-09-8 | |
| 270°C | |
| MFCD00008471 | |
| dimethyl suberate, octanedioic acid, dimethyl ester, suberic acid, dimethyl ester, octanedioic acid dimethyl ester, suberic acid dimethyl ester, octanedioic acid dimethyl, 1,8-dimethyl octanedioate, octanedioic acid, 1,8-dimethyl ester, dimethyl octane-1,8-dioate | |
| COC(=O)CCCCCCC(=O)OC | |
| 202.25 | |
| CHEBI:81345 | |
| ≥98.0% (GC) | |
| Dimethyl Suberate |
| Yellow | |
| C10H18O4 | |
| 25 mL | |
| LNLCRJXCNQABMV-UHFFFAOYSA-N | |
| 1,8-dimethyl octanedioate | |
| 15611 | |
| 202.25 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0879
TSCA : Yes