missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Suberate 98.0+%, TCI America™

Catalog No. O018325ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O018325ML 25 mL
O0183500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O018325ML Supplier TCI America Supplier No. O018325ML

Chemical Identifiers

CAS 1732-09-8
Molecular Formula C10H18O4
Molecular Weight (g/mol) 202.25
MDL Number MFCD00008471
InChI Key LNLCRJXCNQABMV-UHFFFAOYSA-N
Synonym dimethyl suberate, octanedioic acid, dimethyl ester, suberic acid, dimethyl ester, octanedioic acid dimethyl ester, suberic acid dimethyl ester, octanedioic acid dimethyl, 1,8-dimethyl octanedioate, octanedioic acid, 1,8-dimethyl ester, dimethyl octane-1,8-dioate
PubChem CID 15611
ChEBI CHEBI:81345
IUPAC Name 1,8-dimethyl octanedioate
SMILES COC(=O)CCCCCCC(=O)OC

Specifications

Color Yellow
Boiling Point 270°C
Quantity 25 mL
Formula Weight 202.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethyl Suberate
EINECSNumber (2)-0879
TSCA Yes