missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Pimelate 98.0+%, TCI America™

Catalog No. D44765G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D44765G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D44765G Supplier TCI America Supplier No. D44765G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1732-08-7
Molecular Formula C9H16O4
Molecular Weight (g/mol) 188.22
MDL Number MFCD00008470
InChI Key SHWINQXIGSEZAP-UHFFFAOYSA-N
Synonym dimethyl pimelate, heptanedioic acid, dimethyl ester, pimelic acid dimethyl ester, pimelic acid, dimethyl ester, dimethyl 1,7-heptanedioate, 1,7-dimethyl heptanedioate, heptanedioic acid, 1,7-dimethyl ester, unii-ge1pf3049f, heptanedioic acid dimethyl ester, 1ov5vo1
PubChem CID 74416
IUPAC Name 1,7-dimethyl heptanedioate
SMILES COC(=O)CCCCCC(=O)OC

Specifications

Color Yellow
Boiling Point 122°C
Quantity 5 g
Formula Weight 188.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethyl Pimelate
EINECSNumber (2)-0879
TSCA Yes