accessibility menu, dialog, popup

UserName

CAS RN 1732-08-7

IUPAC Name: 1,7-dimethyl heptanedioate
Synonyms: heptanedioic acid, dimethyl ester
Molecular Weight (g/mol): 188.22
Molecular Formula: C9H16O4
InChi Key: SHWINQXIGSEZAP-UHFFFAOYSA-N
SMILES: COC(=O)CCCCCC(=O)OC
Boiling Point 
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)