missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl pimelate, 98+%

Catalog No. p-7026175
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2202606 5 g
AAB2202614 25 g
AAB2202622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2202606 Supplier Thermo Scientific Chemicals Supplier No. B2202606
Only null left

CAS: 1732-08-7 | C9H16O4 | 188.22 g/mol

Dimethyl pimelate was used as the internal standard in the analysis of phthalates, sebacates, epoxidized soybean oil (ESBO) and polyadipates after transesterification to the ethyl esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl pimelate was used as the internal standard in the analysis of phthalates, sebacates, epoxidized soybean oil (ESBO) and polyadipates after transesterification to the ethyl esters.

Solubility
Soluble in ethanol, benzene, ether. Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 1732-08-7
Molecular Formula C9H16O4
Molecular Weight (g/mol) 188.22
MDL Number MFCD00008470
InChI Key SHWINQXIGSEZAP-UHFFFAOYSA-N
Synonym dimethyl pimelate, heptanedioic acid, dimethyl ester, pimelic acid dimethyl ester, pimelic acid, dimethyl ester, dimethyl 1,7-heptanedioate, 1,7-dimethyl heptanedioate, heptanedioic acid, 1,7-dimethyl ester, unii-ge1pf3049f, heptanedioic acid dimethyl ester, 1ov5vo1
PubChem CID 74416
IUPAC Name dimethyl heptanedioate
SMILES COC(=O)CCCCCC(=O)OC

Specifications

Density 1.041
Boiling Point 121°C to 122°C (11 mmHg)
Flash Point >110°C (230°F)
Odor Mild
Refractive Index 1.431
Linear Formula CH3O2C(CH2)5CO2CH3
Quantity 5 g
Beilstein 1777309
Solubility Information Soluble in ethanol,benzene,ether. Slightly soluble in water.
Formula Weight 188.23
Percent Purity 99%
Chemical Name or Material Dimethyl pimelate
Show More Show Less
EINECSNumber 217-057-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only