missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl Octadecanedioate 96.0+%, TCI America™
Supplier: TCI America D354025G
Specifications
| Dimethyl Octadecanedioate | |
| 60°C | |
| 207°C | |
| MFCD00673461 | |
| octadecanedioic acid, dimethyl ester, octadecanedioic acid dimethyl ester, acmc-209cyd, dimethyl 1,18-octadecanedioate, bis-methyl 1,18-octadecanedioate, octadecadioic acid, dimethyl ester, 1,18-dimethyl octadecanedioate, octadecanedioic acid,1,18-dimethyl ester | |
| COC(=O)CCCCCCCCCCCCCCCCC(=O)OC | |
| 342.52 | |
| 342.52 | |
| Crystalline Powder |
| 1472-93-1 | |
| White-Yellow | |
| C20H38O4 | |
| 25 g | |
| ZOXMHKCFDXHCJX-UHFFFAOYSA-N | |
| 1,18-dimethyl octadecanedioate | |
| 566760 | |
| ≥96.0% (GC) |
Chemical Identifiers
| 1472-93-1 | |
| 342.52 | |
| ZOXMHKCFDXHCJX-UHFFFAOYSA-N | |
| 566760 | |
| COC(=O)CCCCCCCCCCCCCCCCC(=O)OC |
| C20H38O4 | |
| MFCD00673461 | |
| octadecanedioic acid, dimethyl ester, octadecanedioic acid dimethyl ester, acmc-209cyd, dimethyl 1,18-octadecanedioate, bis-methyl 1,18-octadecanedioate, octadecadioic acid, dimethyl ester, 1,18-dimethyl octadecanedioate, octadecanedioic acid,1,18-dimethyl ester | |
| 1,18-dimethyl octadecanedioate |
Safety and Handling
EINECSNumber : (2)-2630
TSCA : Yes