missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Octadecanedioate 96.0+%, TCI America™

Catalog No. p-7127779
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D35405G 5 g
D354025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D35405G Supplier TCI America Supplier No. D35405G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1472-93-1
Molecular Formula C20H38O4
Molecular Weight (g/mol) 342.52
MDL Number MFCD00673461
InChI Key ZOXMHKCFDXHCJX-UHFFFAOYSA-N
Synonym octadecanedioic acid, dimethyl ester, octadecanedioic acid dimethyl ester, acmc-209cyd, dimethyl 1,18-octadecanedioate, bis-methyl 1,18-octadecanedioate, octadecadioic acid, dimethyl ester, 1,18-dimethyl octadecanedioate, octadecanedioic acid,1,18-dimethyl ester
PubChem CID 566760
IUPAC Name 1,18-dimethyl octadecanedioate
SMILES COC(=O)CCCCCCCCCCCCCCCCC(=O)OC

Specifications

Melting Point 60°C
Color White-Yellow
Boiling Point 207°C
Quantity 5 g
Formula Weight 342.52
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl Octadecanedioate
EINECSNumber (2)-2630
TSCA Yes