missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dimethyl Nitroterephthalate, TCI America™

$32.08 - $168.06

Chemical Identifiers

CAS 5292-45-5
Molecular Formula C10H9NO6
Molecular Weight (g/mol) 239.183
MDL Number MFCD00008422
InChI Key PAYWCKGMOYQZAW-UHFFFAOYSA-N
Synonym dimethyl 2-nitroterephthalate, dimethyl nitroterephthalate, terephthalic acid, nitro-, dimethyl ester, unii-59qlu09dz4, 1,4-benzenedicarboxylic acid, 2-nitro-, dimethyl ester, 1,4-benzenedicarboxylic acid, 2-nitro-, 1,4-dimethyl ester, nitroterephthalic acid dimethyl ester, 1,4-dimethyl 2-nitrobenzene-1,4-dicarboxylate, acmc-1aj3c, di-methyl nitroterephthalate
PubChem CID 21364
IUPAC Name dimethyl 2-nitrobenzene-1,4-dicarboxylate
SMILES COC(=O)C1=CC(=C(C=C1)C(=O)OC)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N050125G View Documents TCI America
N050125G
25g Each for $32.08
 
N0501250G View Documents TCI America
N0501250G
250g Each for $168.06
 
Specifications

Chemical Identifiers

5292-45-5
239.183
PAYWCKGMOYQZAW-UHFFFAOYSA-N
21364
COC(=O)C1=CC(=C(C=C1)C(=O)OC)[N+](=O)[O-]
C10H9NO6
MFCD00008422
dimethyl 2-nitroterephthalate, dimethyl nitroterephthalate, terephthalic acid, nitro-, dimethyl ester, unii-59qlu09dz4, 1,4-benzenedicarboxylic acid, 2-nitro-, dimethyl ester, 1,4-benzenedicarboxylic acid, 2-nitro-, 1,4-dimethyl ester, nitroterephthalic acid dimethyl ester, 1,4-dimethyl 2-nitrobenzene-1,4-dicarboxylate, acmc-1aj3c, di-methyl nitroterephthalate
dimethyl 2-nitrobenzene-1,4-dicarboxylate

Specifications

White-Yellow
25g
C10H9NO6
dimethyl 2-nitroterephthalate, dimethyl nitroterephthalate, terephthalic acid, nitro-, dimethyl ester, unii-59qlu09dz4, 1,4-benzenedicarboxylic acid, 2-nitro-, dimethyl ester, 1,4-benzenedicarboxylic acid, 2-nitro-, 1,4-dimethyl ester, nitroterephthalic acid dimethyl ester, 1,4-dimethyl 2-nitrobenzene-1,4-dicarboxylate, acmc-1aj3c, di-methyl nitroterephthalate
COC(=O)C1=CC(=C(C=C1)C(=O)OC)[N+](=O)[O-]
239.183
239.18
≥97.0% (GC)
75°C
5292-45-5
MFCD00008422
PAYWCKGMOYQZAW-UHFFFAOYSA-N
dimethyl 2-nitrobenzene-1,4-dicarboxylate
21364
Crystal-Powder at 20°C
Dimethyl Nitroterephthalate
Safety and Handling

Safety and Handling

TSCA : Yes