missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl Nitroterephthalate 97.0+%, TCI America™
Supplier: TCI America N0501250G
Specifications
| Dimethyl Nitroterephthalate | |
| 75°C | |
| C10H9NO6 | |
| 250 g | |
| PAYWCKGMOYQZAW-UHFFFAOYSA-N | |
| 1,4-dimethyl 2-nitrobenzene-1,4-dicarboxylate | |
| 21364 | |
| ≥97.0% (GC) |
| 5292-45-5 | |
| White-Yellow | |
| MFCD00008422 | |
| dimethyl 2-nitroterephthalate, dimethyl nitroterephthalate, terephthalic acid, nitro-, dimethyl ester, unii-59qlu09dz4, 1,4-benzenedicarboxylic acid, 2-nitro-, dimethyl ester, 1,4-benzenedicarboxylic acid, 2-nitro-, 1,4-dimethyl ester, nitroterephthalic acid dimethyl ester, 1,4-dimethyl 2-nitrobenzene-1,4-dicarboxylate, acmc-1aj3c, di-methyl nitroterephthalate | |
| COC(=O)C1=CC=C(C(=O)OC)C(=C1)[N+]([O-])=O | |
| 239.18 | |
| 239.18 | |
| Crystalline Powder |
Chemical Identifiers
| 5292-45-5 | |
| 239.18 | |
| PAYWCKGMOYQZAW-UHFFFAOYSA-N | |
| 21364 | |
| COC(=O)C1=CC=C(C(=O)OC)C(=C1)[N+]([O-])=O |
| C10H9NO6 | |
| MFCD00008422 | |
| dimethyl 2-nitroterephthalate, dimethyl nitroterephthalate, terephthalic acid, nitro-, dimethyl ester, unii-59qlu09dz4, 1,4-benzenedicarboxylic acid, 2-nitro-, dimethyl ester, 1,4-benzenedicarboxylic acid, 2-nitro-, 1,4-dimethyl ester, nitroterephthalic acid dimethyl ester, 1,4-dimethyl 2-nitrobenzene-1,4-dicarboxylate, acmc-1aj3c, di-methyl nitroterephthalate | |
| 1,4-dimethyl 2-nitrobenzene-1,4-dicarboxylate |
Safety and Handling
TSCA : Yes