missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl naphthalene-2,6-dicarboxylate, 99+%

Catalog No. AAL1768906
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1768906 5 g
AAL1768914 25 g
AAL1768922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1768906 Supplier Thermo Scientific Chemicals Supplier No. L1768906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 840-65-3 | C14H12O4 | 244.246 g/mol

Dimethyl naphthalene-2,6-dicarboxylate is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl naphthalene-2,6-dicarboxylate is used as a pharmaceutical intermediate.

Solubility
Soluble in hot toluene (very faint turbidity). Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 840-65-3
Molecular Formula C14H12O4
Molecular Weight (g/mol) 244.246
MDL Number MFCD00004100
InChI Key GYUVMLBYMPKZAZ-UHFFFAOYSA-N
Synonym dimethyl 2,6-naphthalenedicarboxylate, 2,6-dicarbomethoxynaphthalene, 2,6-naphthalenedicarboxylic acid, dimethyl ester, unii-x4q5t8rs0g, dimethyl-2,6-naphthalenedicarboxylate, 2,6-naphthalenedicarboxylic acid dimethyl ester, x4q5t8rs0g, 2,6-naphthalene dicarboxylic acid, dimethyl ester, dimethylnaphthalene-2,6-dicarboxylate, 2,6-naphthalenedicarboxylic acid, 2,6-dimethyl ester
PubChem CID 61225
IUPAC Name dimethyl naphthalene-2,6-dicarboxylate
SMILES COC(=O)C1=CC2=C(C=C1)C=C(C=C2)C(=O)OC

Specifications

Melting Point 187°C to 193°C
Flash Point 232°C (449°F)
Quantity 5 g
Beilstein 987259
Solubility Information Soluble in hot toluene (very faint turbidity). Insoluble in water.
Formula Weight 244.25
Percent Purity ≥99%
Chemical Name or Material Dimethyl naphthalene-2,6-dicarboxylate
EINECSNumber 212-661-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only