accessibility menu, dialog, popup

UserName

CAS RN 840-65-3

IUPAC Name: 2,6-dimethyl naphthalene-2,6-dicarboxylate
Synonyms: dimethyl 2,6-naphthalate
Molecular Weight (g/mol): 244.25
Molecular Formula: C14H12O4
InChi Key: GYUVMLBYMPKZAZ-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC2=CC=C(C=C2C=C1)C(=O)OC
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)