missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Icosanedioate 95.0+%, TCI America™

Supplier:  TCI America E033025G

 View more versions of this product

Catalog No. E033025G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Dimethyl Icosanedioate
66°C
223°C
MFCD00059302
Dimethyl Eicosanedioate, Eicosanedioic Acid Dimethyl Ester, Icosanedioic Acid Dimethyl Ester
COC(=O)CCCCCCCCCCCCCCCCCCC(=O)OC
370.574
370.57
Crystalline Powder
42235-38-1
White
C22H42O4
25 g
PODMJVMUYCFFMK-UHFFFAOYSA-N
dimethyl icosanedioate
566668
≥95.0% (GC)

Chemical Identifiers

42235-38-1
370.574
PODMJVMUYCFFMK-UHFFFAOYSA-N
566668
COC(=O)CCCCCCCCCCCCCCCCCCC(=O)OC
C22H42O4
MFCD00059302
Dimethyl Eicosanedioate, Eicosanedioic Acid Dimethyl Ester, Icosanedioic Acid Dimethyl Ester
dimethyl icosanedioate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions