missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Icosanedioate 95.0+%, TCI America™

Catalog No. p-7128940
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E03305G 5 g
E033025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E03305G Supplier TCI America Supplier No. E03305G

Chemical Identifiers

CAS 42235-38-1
Molecular Formula C22H42O4
Molecular Weight (g/mol) 370.574
MDL Number MFCD00059302
InChI Key PODMJVMUYCFFMK-UHFFFAOYSA-N
Synonym Dimethyl Eicosanedioate, Eicosanedioic Acid Dimethyl Ester, Icosanedioic Acid Dimethyl Ester
PubChem CID 566668
IUPAC Name dimethyl icosanedioate
SMILES COC(=O)CCCCCCCCCCCCCCCCCCC(=O)OC

Specifications

Melting Point 66°C
Color White
Boiling Point 223°C
Quantity 5 g
Formula Weight 370.57
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl Icosanedioate