missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl Hexadecanedioate 95.0+%, TCI America™
Supplier: TCI America D355425G
Specifications
| Dimethyl Hexadecanedioate | |
| 53°C | |
| 172°C | |
| MFCD00039806 | |
| Hexadecanedioic Acid Dimethyl Ester | |
| COC(=O)CCCCCCCCCCCCCCC(=O)OC | |
| 314.47 | |
| 314.47 | |
| Crystalline Powder |
| 19102-90-0 | |
| White-Yellow | |
| C18H34O4 | |
| 25 g | |
| MQIZSSSBYFJIJF-UHFFFAOYSA-N | |
| 1,16-dimethyl hexadecanedioate | |
| 140477 | |
| ≥95.0% (GC) |
Chemical Identifiers
| 19102-90-0 | |
| 314.47 | |
| MQIZSSSBYFJIJF-UHFFFAOYSA-N | |
| 140477 | |
| COC(=O)CCCCCCCCCCCCCCC(=O)OC |
| C18H34O4 | |
| MFCD00039806 | |
| Hexadecanedioic Acid Dimethyl Ester | |
| 1,16-dimethyl hexadecanedioate |
Safety and Handling
EINECSNumber : (2)-2630
TSCA : No