accessibility menu, dialog, popup

UserName

H3COOOOCH3

CAS RN 19102-90-0

IUPAC Name: 1,16-dimethyl hexadecanedioate
Synonyms: 1,16-dimethyl hexadecanedioate
Molecular Weight (g/mol): 314.47
Molecular Formula: C18H34O4
InChi Key: MQIZSSSBYFJIJF-UHFFFAOYSA-N
SMILES: COC(=O)CCCCCCCCCCCCCCC(=O)OC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results