missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Hexadecanedioate 95.0+%, TCI America™

Catalog No. D35545G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D35545G 5 g
D355425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D35545G Supplier TCI America Supplier No. D35545G

Chemical Identifiers

CAS 19102-90-0
Molecular Formula C18H34O4
Molecular Weight (g/mol) 314.47
MDL Number MFCD00039806
InChI Key MQIZSSSBYFJIJF-UHFFFAOYSA-N
Synonym Hexadecanedioic Acid Dimethyl Ester
PubChem CID 140477
IUPAC Name 1,16-dimethyl hexadecanedioate
SMILES COC(=O)CCCCCCCCCCCCCCC(=O)OC

Specifications

Melting Point 53°C
Color White-Yellow
Boiling Point 172°C
Quantity 5 g
Formula Weight 314.47
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl Hexadecanedioate
EINECSNumber (2)-2630
TSCA No