missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl cyanodithioiminocarbonate, 95%

Catalog No. AAA1104314
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1104314 25 g
AAA1104322 100 g
AAA1104336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1104314 Supplier Thermo Scientific Chemicals Supplier No. A1104314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 10191-60-3 | C4H6N2S2 | 146.226 g/mol

Dimethyl cyanodithioiminocarbonate has been used in the synthesis of 4-methylthiopyrazolo[1,5-a]-1,3,5-triazines, methylsulfanylpyrimidines, cyanoguanidines and N-aryl-6-methylsulfanyl-4-oxopyrimidine-5-carbonitriles. It has been also used in the synthesis of methylsulfanyl derivatives of azoloazines and azoloazoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl cyanodithioiminocarbonate has been used in the synthesis of 4-methylthiopyrazolo[1,5-a]-1,3,5-triazines, methylsulfanylpyrimidines, cyanoguanidines and N-aryl-6-methylsulfanyl-4-oxopyrimidine-5-carbonitriles. It has been also used in the synthesis of methylsulfanyl derivatives of azoloazines and azoloazoles.

Solubility
Slightly soluble in methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 10191-60-3
Molecular Formula C4H6N2S2
Molecular Weight (g/mol) 146.226
MDL Number MFCD00009825
InChI Key IULFXBLVJIPESI-UHFFFAOYSA-N
Synonym dimethyl cyanocarbonimidodithioate, dimethyl cyanodithioiminocarbonate, dimethyl n-cyanodithioiminocarbonate, carbonimidodithioic acid, cyano-, dimethyl ester, s,s-dimethyl cyanoimidodithiocarbonate, cyanoimino bis methylthio methane, dimethyl n-cyanoimidodithiocarbonate, dimethyl cyanimidodithiocarbonate, dimethyl cyanoiminodithiocarbonate, dimethyl n-cyanodithiocarbonimidate
PubChem CID 66289
IUPAC Name bis(methylsulfanyl)methylidenecyanamide
SMILES CSC(=NC#N)SC

Specifications

Melting Point 50°C to 56°C
Flash Point >110°C (230°F)
Odor Stench
Linear Formula NCN=C(SCH3)2
Quantity 25 g
UN Number UN2923
Beilstein 635998
Solubility Information Slightly soluble in methanol.
Formula Weight 146.23
Percent Purity 95%
Chemical Name or Material Dimethyl cyanodithioiminocarbonate
Show More Show Less
Hazard Category H301+H311+H331-H314-H335
Hazard Statement GHS H Statement
H314-H318-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
EINECSNumber 233-467-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only