missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl Adipate 99.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 110°C |
| Quantity | 25 mL |
| Formula Weight | 174.20 |
| Percent Purity | ≥99.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Dimethyl Adipate |
Safety and Handling
| EINECSNumber | (2)-0861&(2)-0879 |
| RTECSNumber | AV1645000 |
| TSCA | Yes |