missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Acetylsuccinate 96.0+%, TCI America™

Catalog No. p-7117266
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A114325G 25 g
A1143500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A114325G Supplier TCI America Supplier No. A114325G
Only null left

Chemical Identifiers

CAS 10420-33-4
Molecular Formula C8H12O5
Molecular Weight (g/mol) 188.18
MDL Number MFCD00008448
InChI Key XREKLQOUFWBSFH-UHFFFAOYNA-N
Synonym dimethyl acetylsuccinate, dimethyl 2-acetylsuccinate, 1,4-dimethyl 2-acetylbutanedioate, butanedioic acid, acetyl-, dimethyl ester, acetylsuccinic acid dimethyl ester, dmas, butanedioic acid, 2-acetyl-, 1,4-dimethyl ester, doimethyl acetylsuccinate, acmc-1bwle, dimethyl 2-acetylsuccite
PubChem CID 112039
IUPAC Name 1,4-dimethyl 2-acetylbutanedioate
SMILES COC(=O)CC(C(C)=O)C(=O)OC

Specifications

Color White
Boiling Point 200°C
Quantity 25 g
Formula Weight 188.18
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl Acetylsuccinate