accessibility menu, dialog, popup

UserName

CAS RN 10420-33-4

IUPAC Name: 1,4-dimethyl 2-acetylbutanedioate
Synonyms: 1,4-dimethyl 2-acetylbutanedioate
Molecular Weight (g/mol): 188.18
Molecular Formula: C8H12O5
InChi Key: XREKLQOUFWBSFH-UHFFFAOYNA-N
SMILES: COC(=O)CC(C(C)=O)C(=O)OC
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)