missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl acetylsuccinate, 96%

Catalog No. AAB2542322
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2542322 100 g
AAB2542336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2542322 Supplier Thermo Scientific Chemicals Supplier No. B2542322
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10420-33-4 | C8H12O5 | 188.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10420-33-4
Molecular Formula C8H12O5
Molecular Weight (g/mol) 188.18
MDL Number MFCD00008448
InChI Key XREKLQOUFWBSFH-UHFFFAOYNA-N
Synonym dimethyl acetylsuccinate, dimethyl 2-acetylsuccinate, 1,4-dimethyl 2-acetylbutanedioate, butanedioic acid, acetyl-, dimethyl ester, acetylsuccinic acid dimethyl ester, dmas, butanedioic acid, 2-acetyl-, 1,4-dimethyl ester, doimethyl acetylsuccinate, acmc-1bwle, dimethyl 2-acetylsuccite
PubChem CID 112039
IUPAC Name dimethyl 2-acetylbutanedioate
SMILES COC(=O)CC(C(C)=O)C(=O)OC

Specifications

Melting Point 30°C to 33°C
Density 1.16
Boiling Point 129°C to 134°C (12 mmHg)
Flash Point >110°C (230°F)
Quantity 100 g
Formula Weight 188.18
Percent Purity 96%
Chemical Name or Material Dimethyl acetylsuccinate
EINECSNumber 233-897-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only