missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Acetylenedicarboxylate 96.0+%, TCI America™

Supplier:  TCI America A0090500ML

 View more versions of this product

Catalog No. A0090500ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Dimethyl Acetylenedicarboxylate
Yellow
C6H6O4
500 mL
dimethyl acetylenedicarboxylate, 2-butynedioic acid, dimethyl ester, 1,4-dimethyl but-2-ynedioate, dimethyl butynedioate, dimethyl 2-butynedioate, acetylenedicarboxylic acid dimethyl ester, dmad, methyl acetylenedicarboxylate, bis methoxycarbonyl acetylene, di carbomethoxy acetylene
COC(=O)C#CC(=O)OC
142.11
142.11
Liquid
762-42-5
198°C
MFCD00008456
1760
VHILMKFSCRWWIJ-UHFFFAOYSA-N
dimethyl but-2-ynedioate
12980
≥96.0% (GC)

Chemical Identifiers

762-42-5
142.11
VHILMKFSCRWWIJ-UHFFFAOYSA-N
12980
COC(=O)C#CC(=O)OC
C6H6O4
MFCD00008456
dimethyl acetylenedicarboxylate, 2-butynedioic acid, dimethyl ester, 1,4-dimethyl but-2-ynedioate, dimethyl butynedioate, dimethyl 2-butynedioate, acetylenedicarboxylic acid dimethyl ester, dmad, methyl acetylenedicarboxylate, bis methoxycarbonyl acetylene, di carbomethoxy acetylene
dimethyl but-2-ynedioate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : ES0175000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions