missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Acetylenedicarboxylate 96.0+%, TCI America™

Catalog No. p-7116707
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A009025ML 25 mL
A0090100ML 100 mL
A0090500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A009025ML Supplier TCI America Supplier No. A009025ML
Only null left

Chemical Identifiers

CAS 762-42-5
Molecular Formula C6H6O4
Molecular Weight (g/mol) 142.11
MDL Number MFCD00008456
InChI Key VHILMKFSCRWWIJ-UHFFFAOYSA-N
Synonym dimethyl acetylenedicarboxylate, 2-butynedioic acid, dimethyl ester, 1,4-dimethyl but-2-ynedioate, dimethyl butynedioate, dimethyl 2-butynedioate, acetylenedicarboxylic acid dimethyl ester, dmad, methyl acetylenedicarboxylate, bis methoxycarbonyl acetylene, di carbomethoxy acetylene
PubChem CID 12980
IUPAC Name dimethyl but-2-ynedioate
SMILES COC(=O)C#CC(=O)OC

Specifications

Color Yellow
Boiling Point 198°C
Quantity 25 mL
UN Number 1760
Formula Weight 142.11
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethyl Acetylenedicarboxylate
RTECSNumber ES0175000
TSCA Yes