missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl acetone-1,3-dicarboxylate, 97%

CAS: 1830-54-2 | C7H10O5 | 174.15 g/mol

Supplier:  Thermo Scientific Chemicals A1496922

Quantity 100 g

 View more versions of this product

Catalog No. AAA1496922

Alert:

This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.


Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1830-54-2
174.15
RNJOKCPFLQMDEC-UHFFFAOYSA-N
74591
C7H10O5
MFCD00008462
dimethyl 1,3-acetonedicarboxylate, dimethyl acetone-1,3-dicarboxylate, dimethyl 3-oxoglutarate, 1,5-dimethyl 3-oxopentanedioate, pentanedioic acid, 3-oxo-, dimethyl ester, dimethyl acetonedicarboxylate, acetone dicarboxylic acid, dimethyl ester, glutaric acid, 3-oxo-, dimethyl ester, pentanedioic acid, 3-oxo-, 1,5-dimethyl ester, dimethyl 3-oxopentane-1,5-dioate
COC(=O)CC(=O)CC(=O)OC

Specifications

Dimethyl acetone-1,3-dicarboxylate
16°C to 17°C
237°C to 241°C
Ester-like
1.444
MFCD00008462
dimethyl 1,3-acetonedicarboxylate, dimethyl acetone-1,3-dicarboxylate, dimethyl 3-oxoglutarate, 1,5-dimethyl 3-oxopentanedioate, pentanedioic acid, 3-oxo-, dimethyl ester, dimethyl acetonedicarboxylate, acetone dicarboxylic acid, dimethyl ester, glutaric acid, 3-oxo-, dimethyl ester, pentanedioic acid, 3-oxo-, 1,5-dimethyl ester, dimethyl 3-oxopentane-1,5-dioate
COC(=O)CC(=O)CC(=O)OC
174.15
174.16
1830-54-2
1.185
>110°C (230°F)
C7H10O5
100 g
1366042
RNJOKCPFLQMDEC-UHFFFAOYSA-N
dimethyl 3-oxopentanedioate
74591
97%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H317
May cause an allergic skin reaction.

P261-P272-P280g-P302+P352-P333+P313-P363-P501c

H317

EINECSNumber : 217-385-8

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions