missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl acetone-1,3-dicarboxylate, 97%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 16°C to 17°C |
| Density | 1.185 |
| Boiling Point | 237°C to 241°C |
| Flash Point | >110°C (230°F) |
| Odor | Ester-like |
| Refractive Index | 1.444 |
| Quantity | 25 g |
| Beilstein | 1366042 |
| IUPAC Name | 1,5-dimethyl 3-oxopentanedioate |
| Formula Weight | 174.16 |
| Show More |
Safety and Handling
| missing translation for 'healthHazard1' | H317 |
| missing translation for 'healthHazard2' | GHS H Statement H317 May cause an allergic skin reaction. |
| missing translation for 'healthHazard3' | P261-P272-P280g-P302+P352-P333+P313-P363-P501c |
| EINECSNumber | 217-385-8 |
| TSCA | Yes |
| Recommended Storage | Keep cold |
RUO – Research Use Only