missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl 5-Aminoisophthalate 98.0+%, TCI America™

Catalog No. p-7117534
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A158625G 25 g
A1586250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A158625G Supplier TCI America Supplier No. A158625G
Only null left

Chemical Identifiers

CAS 99-27-4
Molecular Formula C10H11NO4
Molecular Weight (g/mol) 209.201
MDL Number MFCD00008435
InChI Key DEKPYXUDJRABNK-UHFFFAOYSA-N
PubChem CID 66831
IUPAC Name dimethyl 5-aminobenzene-1,3-dicarboxylate
SMILES COC(=O)C1=CC(=CC(=C1)N)C(=O)OC

Specifications

Melting Point 181°C
Quantity 25 g
Synonym dimethyl 5-aminoisophthalate, 3,5-dicarbomethoxyaniline, 5-aminoisophthalic acid dimethyl ester, 1,3-benzenedicarboxylic acid, 5-amino-, dimethyl ester, 1,3-dimethyl 5-aminobenzene-1,3-dicarboxylate, dimethyl5-aminoisophthalate, 5-amino-isophthalic acid dimethyl ester, 5-amino-dimethyl iso-phthalate, 5-amino-dimethylisophthalate, dimethyl-5-aminoisophthalate
Formula Weight 209.20
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl 5-Aminoisophthalate
EINECSNumber (3)-3026
TSCA Yes