accessibility menu, dialog, popup

UserName

CH3OONH2OOH3C

CAS RN 99-27-4

IUPAC Name: 1,3-dimethyl 5-aminobenzene-1,3-dicarboxylate
Synonyms: 1,3-dimethyl 5-aminobenzene-1,3-dicarboxylate
Molecular Weight (g/mol): 209.2
Molecular Formula: C10H11NO4
InChi Key: DEKPYXUDJRABNK-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(=CC(N)=C1)C(=O)OC
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)