missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl (4-fluorobenzyl)malonate, 97%
CAS: 252938-22-0 | C12H13FO4 | 240.23 g/mol
$89.15 - $254.21
Chemical Identifiers
| CAS | 252938-22-0 |
|---|---|
| Molecular Formula | C12H13FO4 |
| Molecular Weight (g/mol) | 240.23 |
| MDL Number | MFCD01934558 |
| InChI Key | WDFUURBBGZTSIA-UHFFFAOYSA-N |
| Synonym | dimethyl 4-fluorobenzyl malonate, dimethyl 2-4-fluorobenzyl malonate, 1,3-dimethyl 2-4-fluorophenyl methyl propanedioate, acmc-1cpd2, dimethyl 4-fluorobenzylmalonate, 2-4-fluorobenzyl-malonic acid dimethyl ester, dimethyl 2-4-fluorophenyl methyl propanedioate, dimethyl 2-4-fluorophenyl methyl propane-1,3-dioate |
| PubChem CID | 2781576 |
| SMILES | COC(=O)C(CC1=CC=C(F)C=C1)C(=O)OC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH2631003
|
Thermo Scientific Chemicals
H2631003 |
1 g |
Each for $89.15
|
|
|||||
|
AAH2631006
|
Thermo Scientific Chemicals
H2631006 |
5 g |
Each for $254.21
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 252938-22-0 | |
| 240.23 | |
| WDFUURBBGZTSIA-UHFFFAOYSA-N | |
| 2781576 |
| C12H13FO4 | |
| MFCD01934558 | |
| dimethyl 4-fluorobenzyl malonate, dimethyl 2-4-fluorobenzyl malonate, 1,3-dimethyl 2-4-fluorophenyl methyl propanedioate, acmc-1cpd2, dimethyl 4-fluorobenzylmalonate, 2-4-fluorobenzyl-malonic acid dimethyl ester, dimethyl 2-4-fluorophenyl methyl propanedioate, dimethyl 2-4-fluorophenyl methyl propane-1,3-dioate | |
| COC(=O)C(CC1=CC=C(F)C=C1)C(=O)OC |
Specifications
| 252938-22-0 | |
| MFCD01934558 | |
| dimethyl 4-fluorobenzyl malonate, dimethyl 2-4-fluorobenzyl malonate, 1,3-dimethyl 2-4-fluorophenyl methyl propanedioate, acmc-1cpd2, dimethyl 4-fluorobenzylmalonate, 2-4-fluorobenzyl-malonic acid dimethyl ester, dimethyl 2-4-fluorophenyl methyl propanedioate, dimethyl 2-4-fluorophenyl methyl propane-1,3-dioate | |
| COC(=O)C(CC1=CC=C(F)C=C1)C(=O)OC | |
| 240.23 | |
| 240.23 | |
| Dimethyl (4-fluorobenzyl)malonate |
| C12H13FO4 | |
| 1 g | |
| WDFUURBBGZTSIA-UHFFFAOYSA-N | |
| 1,3-dimethyl 2-[(4-fluorophenyl)methyl]propanedioate | |
| 2781576 | |
| 97% |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only