missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl (4-fluorobenzyl)malonate, 97%

Catalog No. AAH2631003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2631003 1 g
AAH2631006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2631003 Supplier Thermo Scientific Chemicals Supplier No. H2631003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 252938-22-0 | C12H13FO4 | 240.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 252938-22-0
Molecular Formula C12H13FO4
Molecular Weight (g/mol) 240.23
MDL Number MFCD01934558
InChI Key WDFUURBBGZTSIA-UHFFFAOYSA-N
Synonym dimethyl 4-fluorobenzyl malonate, dimethyl 2-4-fluorobenzyl malonate, 1,3-dimethyl 2-4-fluorophenyl methyl propanedioate, acmc-1cpd2, dimethyl 4-fluorobenzylmalonate, 2-4-fluorobenzyl-malonic acid dimethyl ester, dimethyl 2-4-fluorophenyl methyl propanedioate, dimethyl 2-4-fluorophenyl methyl propane-1,3-dioate
PubChem CID 2781576
SMILES COC(=O)C(CC1=CC=C(F)C=C1)C(=O)OC

Specifications

Quantity 1 g
IUPAC Name 1,3-dimethyl 2-[(4-fluorophenyl)methyl]propanedioate
Formula Weight 240.23
Percent Purity 97%
Chemical Name or Material Dimethyl (4-fluorobenzyl)malonate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only