missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl 4-Acetoxyisophthalate 98.0+%, TCI America™

$213.69 - $741.20

Chemical Identifiers

CAS 71932-29-1
Molecular Formula C12H12O6
Molecular Weight (g/mol) 252.222
MDL Number MFCD00143241
InChI Key MZBAKPXGINZGLK-UHFFFAOYSA-N
Synonym 4-Acetoxyisophthalic Acid Dimethyl Ester
PubChem CID 23512524
IUPAC Name dimethyl 4-acetyloxybenzene-1,3-dicarboxylate
SMILES CC(=O)OC1=C(C=C(C=C1)C(=O)OC)C(=O)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A12261G
View Documents
TCI America
A12261G
1 g
Each for $213.69
Only null left
 
A12265G
View Documents
TCI America
A12265G
5 g
Each for $741.20
Only null left
 
Description

Description

Specifications

Chemical Identifiers

71932-29-1
252.222
MZBAKPXGINZGLK-UHFFFAOYSA-N
23512524
CC(=O)OC1=C(C=C(C=C1)C(=O)OC)C(=O)OC
C12H12O6
MFCD00143241
4-Acetoxyisophthalic Acid Dimethyl Ester
dimethyl 4-acetyloxybenzene-1,3-dicarboxylate

Specifications

71932-29-1
White
MFCD00143241
4-Acetoxyisophthalic Acid Dimethyl Ester
CC(=O)OC1=C(C=C(C=C1)C(=O)OC)C(=O)OC
252.222
252.22
Crystalline Powder
96°C
C12H12O6
1 g
MZBAKPXGINZGLK-UHFFFAOYSA-N
dimethyl 4-acetyloxybenzene-1,3-dicarboxylate
23512524
≥98.0% (GC)
Dimethyl 4-Acetoxyisophthalate
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'