missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl 4-Acetoxyisophthalate 98.0+%, TCI America™

Catalog No. A12265G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A12265G 5 g
A12261G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A12265G Supplier TCI America Supplier No. A12265G
Only null left

Chemical Identifiers

CAS 71932-29-1
Molecular Formula C12H12O6
Molecular Weight (g/mol) 252.222
MDL Number MFCD00143241
InChI Key MZBAKPXGINZGLK-UHFFFAOYSA-N
Synonym 4-Acetoxyisophthalic Acid Dimethyl Ester
PubChem CID 23512524
IUPAC Name dimethyl 4-acetyloxybenzene-1,3-dicarboxylate
SMILES CC(=O)OC1=C(C=C(C=C1)C(=O)OC)C(=O)OC

Specifications

Melting Point 96°C
Color White
Quantity 5 g
Formula Weight 252.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl 4-Acetoxyisophthalate