missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl 2-Oxoglutarate 95.0+%, TCI America™

Supplier:  TCI America K001325ML

Quantity 25 mL

 View more versions of this product

Catalog No. K001325ML

Only null left

Description

Description

Specifications

Chemical Identifiers

13192-04-6
174.152
TXIXSLPEABAEHP-UHFFFAOYSA-N
25775
COC(=O)CCC(=O)C(=O)OC
C7H10O5
MFCD00048052
Dimethyl 2-Ketoglutarate, 2-Ketoglutaric Acid Dimethyl Ester, 2-Oxoglutaric Acid Dimethyl Ester
dimethyl 2-oxopentanedioate

Specifications

Dimethyl 2-Oxoglutarate
Yellow
C7H10O5
MFCD00048052
TXIXSLPEABAEHP-UHFFFAOYSA-N
dimethyl 2-oxopentanedioate
25775
≥95.0% (GC)
13192-04-6
117°C
25 mL
Dimethyl 2-Ketoglutarate, 2-Ketoglutaric Acid Dimethyl Ester, 2-Oxoglutaric Acid Dimethyl Ester
COC(=O)CCC(=O)C(=O)OC
174.152
174.15
Liquid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions