missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl 2-Oxoglutarate 95.0+%, TCI America™

Catalog No. p-7131741
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K00135ML 5 mL
K001325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. K00135ML Supplier TCI America Supplier No. K00135ML
Only null left

Chemical Identifiers

CAS 13192-04-6
Molecular Formula C7H10O5
Molecular Weight (g/mol) 174.152
MDL Number MFCD00048052
InChI Key TXIXSLPEABAEHP-UHFFFAOYSA-N
Synonym Dimethyl 2-Ketoglutarate, 2-Ketoglutaric Acid Dimethyl Ester, 2-Oxoglutaric Acid Dimethyl Ester
PubChem CID 25775
IUPAC Name dimethyl 2-oxopentanedioate
SMILES COC(=O)CCC(=O)C(=O)OC

Specifications

Color Yellow
Boiling Point 117°C
Quantity 5 mL
Formula Weight 174.15
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethyl 2-Oxoglutarate