missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl 2,7-Naphthalenedicarboxylate 98.0+%, TCI America™

Catalog No. N06841G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N06841G 1 g
N06845G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N06841G Supplier TCI America Supplier No. N06841G

Chemical Identifiers

CAS 2549-47-5
Molecular Formula C14H12O4
Molecular Weight (g/mol) 244.246
MDL Number MFCD03427560
InChI Key WYIBAMPRACRCOM-UHFFFAOYSA-N
Synonym 2,7-Naphthalenedicarboxylic Acid Dimethyl Ester
PubChem CID 9881315
IUPAC Name dimethyl naphthalene-2,7-dicarboxylate
SMILES COC(=O)C1=CC2=C(C=C1)C=CC(=C2)C(=O)OC

Specifications

Melting Point 138°C
Color White-Yellow
Quantity 1 g
Formula Weight 244.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl 2,7-Naphthalenedicarboxylate