accessibility menu, dialog, popup

UserName

H3COOOOCH3

CAS RN 2549-47-5

IUPAC Name: 2,7-dimethyl naphthalene-2,7-dicarboxylate
Synonyms: 2,7-dimethyl naphthalene-2,7-dicarboxylate
Molecular Weight (g/mol): 244.25
Molecular Formula: C14H12O4
InChi Key: WYIBAMPRACRCOM-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC2=CC(=CC=C2C=C1)C(=O)OC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results