missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl 2,6-Naphthalenedicarboxylate 99.0+%, TCI America™

Catalog No. p-7133773
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
N03645G 5 g
N036425G 25 g
2 options

Catalog No. N03645G

Supplier: TCI America N03645G

Only null left

Chemical Identifiers

CAS 840-65-3
Molecular Formula C14H12O4
Molecular Weight (g/mol) 244.246
MDL Number MFCD00004100
InChI Key GYUVMLBYMPKZAZ-UHFFFAOYSA-N
Synonym dimethyl 2,6-naphthalenedicarboxylate, 2,6-dicarbomethoxynaphthalene, 2,6-naphthalenedicarboxylic acid, dimethyl ester, unii-x4q5t8rs0g, dimethyl-2,6-naphthalenedicarboxylate, 2,6-naphthalenedicarboxylic acid dimethyl ester, x4q5t8rs0g, 2,6-naphthalene dicarboxylic acid, dimethyl ester, dimethylnaphthalene-2,6-dicarboxylate, 2,6-naphthalenedicarboxylic acid, 2,6-dimethyl ester
PubChem CID 61225
IUPAC Name dimethyl naphthalene-2,6-dicarboxylate
SMILES COC(=O)C1=CC2=C(C=C1)C=C(C=C2)C(=O)OC

Specifications

Melting Point 192°C
Color White-Yellow
Quantity 5 g
Formula Weight 244.25
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl 2,6-Naphthalenedicarboxylate
EINECSNumber (4)-0409
TSCA Yes